Reagent Reservoir Assembly - FAQs
Reagent Reservoir Assembly - FAQs
View additional product information for Reagent Reservoir Assembly - FAQs (N07489)
1 product FAQs found
Can the Multidrop Combi nL reagent reservoir be autoclaved?
Yes, the reagent reservoir can be autoclaved.