REAGENT RESERVOIR 1000 ML NON - FAQs
REAGENT RESERVOIR 1000 ML NON - FAQs
View additional product information for REAGENT RESERVOIR 1000 ML NON - FAQs (N07490)
1 product FAQs found
Can the Multidrop Combi nL reagent reservoir be autoclaved?
Yes, the reagent reservoir can be autoclaved.