GLASS REAGENT RESERVOIR 1000ML - FAQs
GLASS REAGENT RESERVOIR 1000ML - FAQs
View additional product information for GLASS REAGENT RESERVOIR 1000ML - FAQs (N09505)
1 product FAQs found
Can the Multidrop Combi nL reagent reservoir be autoclaved?
Yes, the reagent reservoir can be autoclaved.