Neon™ NxT Electroporation System 100 μL Kit with 8-Channel Tubes
Neon™ NxT Electroporation System 100 μL Kit with 8-Channel Tubes
Neon™ NxT Electroporation System 100 μL Kit with 8-Channel Tubes
Neon™ NxT Electroporation System 100 μL Kit with 8-Channel Tubes
Neon™ NxT Electroporation System 100 μL Kit with 8-Channel Tubes
Invitrogen™

Neon™ NxT Electroporation System 100 μL Kit with 8-Channel Tubes

The Neon NxT Electroporation System 100-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System with 8-channel pipette and pipette station.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
N100384-8384 x 2 Reactions
N10096-896 x 2 Reactions
Catalog number N100384-8
Price (USD)
7,029.65
Online exclusive
7,400.00
Save 370.35
Each
No. of Reactions:
384 x 2 Reactions
Price (USD)
7,029.65
Online exclusive
7,400.00
Save 370.35
Each

The Neon NxT Electroporation System 100-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System with 8-channel pipette and pipette station (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Use this kit for transfection volumes of 100 μL, containing 5 x 105 to 2 x 106 adherent cells or 1 x 106 to 5 x 106 suspension cells. Cells that have been transfected using the included 100-μL Neon NxT tips are then ready to be plated into a multi-well culture dish.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Transfection
For Use With (Equipment)Neon™ NxT Electroporation System
No. of Reactions384 x 2 Reactions
Quantity384 x 2 Reactions
Volume (Metric) Sample100 μL
Cell TypeStem Cells, Primary Cells, Hard-to-Transfect Cells
FormatElectroporation Pipette Tip
Transfection TechniqueElectroporation
Unit SizeEach
Contents & Storage
Box 1 includes:
• Resuspension R Buffer, 2 x 80 mL
• Resuspension T Buffer, 2 x 80 mL
• Resuspension Genome Editing Buffer, 2 x 80 mL
• Electrolytic E100 Buffer, 2 x 100 mL

Store at 4°C.

Box 2 includes:
• Reaction tips, 384 tips
• Electroporation 8-channel tubes, 16 tubes

Store at room temperature.