Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers
Thermo Scientific™
Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers
Enhance your Wellwash and Wellwash Versa microplate washers with a range of accessories and replacement parts. Wellwash microplate washers enable reliable ELISA plate washing for 96- or 384-well plates with low residual wash volumes and user-friendly features for optimal convenience and safety.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
N10806Waste BottleWellwash Microplate Strip Washer
N10805Wash BottleWellwash Microplate Strip Washer
N10815Aerosol CoverWellwash Microplate Strip Washer
N10817Bottle StandWellwash Microplate Strip Washer
N10800Wash Head (1 x 8)Wellwash Microplate Strip Washer
N10801Wash Head (1 x 12)Wellwash Microplate Strip Washer
N10802Wash Head (2 x 8)Wellwash Versa Microplate Strip Washer
N10803Wash Head (1 x 16)Wellwash Versa Microplate Strip Washer
N10804Wash Head (Blank)Wellwash Versa Microplate Strip Washer
N10807Wash Bottle AWellwash Versa Microplate Strip Washer
N10808Wash Bottle BWellwash Versa Microplate Strip Washer
N10809Rinse BottleWellwash Versa Microplate Strip Washer
N10810Waste BottleWellwash Microplate Strip Washer
N10811Wash Bottle AWellwash Microplate Strip Washer
N10812Wash Bottle BWellwash Versa Microplate Strip Washer
N10813Rinse BottleWellwash Versa Microplate Strip Washer
N10814Waste BottleWellwash Versa Microplate Strip Washer
N10816Aerosol CoverWellwash Versa Microplate Strip Washer
N10818Bottle Stand, 2 x 2 ConfigurationWellwash Versa Microplate Strip Washer
N10819Bottle Stand, 1 x 4 ConfigurationWellwash Versa Microplate Strip Washer
N10820Bottle StandWellwash Microplate Strip Washer
N10821Storage BottleWellwash Versa Microplate Strip Washer
N12407Liquid FilterWellwash Microplate Strip Washer
N12785Storage Bottle CapWellwash Microplate Strip Washer
Catalog number N10806
Price (USD)
402.00
Each
Product Type:
Waste Bottle
For Use With (Equipment):
Wellwash Microplate Strip Washer
Price (USD)
402.00
Each
These accessories and replacement parts are for use with the Wellwash and Wellwash Versa microplate washers. Some are compatible with both models, while others are specifically designed for either the Wellwash or Wellwash Versa microplate washer.
Wellwash microplate strip washers provide reliable and robust plate washing for ELISA applications with low residual wash volumes. Wellwash microplate washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease of use, convenience, and safety.
Specifications
Capacity (Metric)2 L
IncludesCap with Tubing and Liquid Level Sensors
Quantity1 Ea.
For Use With (Equipment)Wellwash Microplate Strip Washer
Product TypeWaste Bottle
Unit SizeEach