Product Image
Thermo Scientific™

KingFisher Duo Prime Accessories

The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormat
N1245996 Deep-well Head
N1246024 Deep-well Head
N1246224 Deep-well Heating Block
N1246196 Deep-well Heating Block
Catalog number N12459
Price (USD)
656.00
Each
Format:
96 Deep-well Head
Price (USD)
656.00
Each
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality.
The KingFisher Duo Prime instrument comes with a 12-pin magnet head for 96 deep well applications, a 6-pin magnet head for 24 deep well applications, and heating blocks. Additional magnet heads and heating blocks are offered here for separate purchase.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DisposableReusable
For Use With (Equipment)KingFisher Duo Prime Purification System
Quantity1 Head
Capacity12 Samples
Format96 Deep-well Head
Product TypeNucleic Acid Extraction and Cell Separation Instrument
Sample TypeFresh or Frozen Plant Tissues, FFPE Tissue, Blood, Cells, Plasma
Volume (Metric) Processing50 to 1000 μL
Unit SizeEach