Multidrop™ Combi Accessories - FAQs
Multidrop™ Combi Accessories - FAQs
View additional product information for Multidrop™ Combi Accessories - FAQs (N12928)
1 product FAQs found
When should I use the Multidrop Combi tube reagent filter accessories for Multidrop Combi small tube cassettes?
We recommend using the filters to help prevent cassette tips from getting clogged during use. Dust, particles, and fibers commonly found in the lab can cause clogging in the Thermo Scientific Multidrop Combi cassettes, preventing reliable dispensing. Keeping particles from entering the tubing helps reduce the likelihood of a clogged tip in small tubing dispensing cassettes. The filter also acts as a weight to hold the tubing in the source vessel. To use the filter, simply remove the tubing weight from the end of the small cassette tubing and replace it with a filter.