Barcode Reader for KingFisher™ Duo Prime
Thermo Scientific™

Barcode Reader for KingFisher™ Duo Prime

Track samples and reagents quickly and easily with the Barcode Reader for Thermo Scientific KingFisher Duo Prime.
Have Questions?
Catalog NumberQuantity
N16640Each
Catalog number N16640
Price (USD)
940.00
Each
Quantity:
Each
Price (USD)
940.00
Each
The Barcode Reader for Thermo Scientific KingFisher Duo Prime is an optional accessory that allows users to track samples directly in the instrument’s internal software quickly and easily.
The Barcode Reader for Thermo Scientific KingFisher Duo Prime is an optional accessory that allows users to track samples directly in the instrument’s internal software quickly and easily.
For research use only. Not for use in diagnostic procedures.
Specifications
Description2D Barcode reader for KingFisher
For Use With (Equipment)KingFisher Duo Prime
QuantityEach
Format1D and 2D
Unit SizeEach

Frequently asked questions (FAQs)

Are the Matrix 500 microliter ScrewTop Tubes (Cat. No. 3743AMB) compatible with the Kingfisher Duo Prime barcode reader?

Yes, the KingFisher Duo Prime barcode reader is able to read the 2D barcode on the Matrix 500 µL ScrewTop Tubes. The barcode reader can generate an error if the tubes are in the tube rack and the barcode position is not able to be scanned. If this occurs, we recommend scanning each tube individually.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.