Poly-HRP Streptavidin - FAQs
Poly-HRP Streptavidin - FAQs
View additional product information for Poly-HRP Streptavidin - FAQs (N200)
2 product FAQs found
What is the concentration of Poly-HRP Streptavidin (Cat. No. N200) and what is the labelling ratio of HRP:Streptavidin?
The concentration of Poly-HRP Streptavidin is 1 mg/mL. The HRP:Streptavidin ratio is 5:1.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center
Can Poly-HRP Streptavidin (Cat. No. N200) be stored at -80 degrees C to increase its shelf life?
We do not recommend storing Poly-HRP Streptavidin at -80 degrees C, as the product can freeze solid at temperatures below -25 degrees C even if it has 50% glycerol as a stabilizer, thereby necessitating thawing of the product. With multiple freeze-thaws, the enzyme conjugates are rendered unstable.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center