F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, NANOGOLD™ - FAQs
F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, NANOGOLD™ - FAQs
View additional product information for F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, NANOGOLD™ - FAQs (N24916)
1 product FAQs found
I want to label F-actin with phalloidin and image with transmission electron microscopy. What is the best way to go about it?
One way is to use a biotin-phalloidin conjugate, then label that with a colloidal gold conjugate of streptavidin. You will need to block endogenous biotin first. Another method is to label with fluorescein phalloidin, then with an anti-fluorescein antibody conjugated with colloidal gold.