Product Image
Thermo Scientific™

Ultra Streptavidin-HRP

Streptavidin-HRP consists of streptavidin protein that is covalently conjugated to horseradish peroxidase (HRP) enzyme. Streptavidin binds to biotin and theRead more
Have Questions?
Catalog NumberQuantity
N5041.5 mL
Catalog number N504
Price (USD)
179.00
Each
Quantity:
1.5 mL
Request bulk or custom format
Price (USD)
179.00
Each
Streptavidin-HRP consists of streptavidin protein that is covalently conjugated to horseradish peroxidase (HRP) enzyme. Streptavidin binds to biotin and the conjugated HRP provides a high level of activity for detection using an appropriate substrate system.

Features of Streptavidin-HRP:

• Product Number: N504
• Size: 1.5 mL
• Formulation: Conjugate in stabilizer solution containing protein and preserved with 0.02% methyliso-thiazolone, 0.02% bromonitrodioxane, and 0.002% other active isothiazolones.
• Storage: Shipped on cold packs. Store at 2-8°C. Stable until expiration date if stored under these conditions.
• Application: ELISA: to be used with biotin-labeled detection antibodies. Recommended dilution range is 1:400 - 1:2,000 in ELISA diluent (e.g., PBS containing 4% protease-free BSA) . The optimal dilution must be determined empirically for each specific application. Prepare dilutions immediately before use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeHRP (Horseradish Peroxidase)
Product TypeStreptavidin Conjugate
Quantity1.5 mL
Target MoleculeBiotin
ConjugateHRP
FormLiquid
Unit SizeEach