What is the biosafety level (BSL) for Human Astrocytes (Cat. No. N7805100)?
The biosafety level (BSL) for mammalian cells in the US is typically biosafety level 2. Please note that this can vary from country to country.
Search
View additional product information for Human Astrocytes - FAQs (N7805100)
1 product FAQs found