MicroAmp™ Cap Strips are designed to fit on MicroAmp™ reaction tubes, tube strips, and 96-well plates. They are available inRead more
Have Questions?
Change view
Catalog Number
Quantity
N8011535
1500 Strips (5 pk./300)
N8010535
300 Strips
2 Options
Catalog number N8011535
Price (USD)
538.65
Online exclusive
586.00
Save 47.35
Each
Quantity:
1500 Strips (5 pk./300)
Price (USD)
538.65
Online exclusive
586.00
Save 47.35
Each
MicroAmp™ Cap Strips are designed to fit on MicroAmp™ reaction tubes, tube strips, and 96-well plates. They are available in 8-cap and 12-cap strips in clear or an assorted set of red, orange, blue, and green. They have a domed profile and are suitable for end-point PCR. For real-time PCR applications, please see our MicroAmp™ Optical 8-Cap Strips (cat. no. 4323032), which have a flat-top profile. Attach the caps using the MicroAmp™ Cap Installing Tool.
Features:
• Will not interfere with sample reading • Provide tight seal to minimize evaporation • Are easy to apply and remove
Minimize Evaporation Use MicroAmp™ Caps Strips to seal the tubes containing your precious samples. This will reduce the possibility of contamination among sample wells and help ensure reliable results.
Easy to Use Use the MicroAmp™ Cap Installing Tool to push the caps down and form a tight seal. When ready, just peel away the caps using the tabs at the end.
Colored for Organization These caps are available in different colors to help you keep track of your tubes during assay preparation.
Specifications
Certifications/ComplianceCertified DNA/Rnase Free
For Use With (Equipment)Thermal Cyclers
Format1,500 Strip Caps
No. of Wells96
Product LineMicroAmp™
Quantity1500 Strips (5 pk./300)
Closure TypeDomed Cap
ColorNatural
MaterialPolypropylene
Product Type8-Strip Cap
Unit SizeEach
Contents & Storage
Includes: 5 packs of N8010535 (Qty 300 per pack)
Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates. Natural (clear) color.