Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
MicroAmp™ Reaction Tube with Cap, 0.2 mL
Applied Biosystems™

MicroAmp™ Reaction Tube with Cap, 0.2 mL

The Applied Biosystems™ MicroAmp™ Reaction Tubes with Caps are optimally designed for precise PCR and are designed for use withRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
N801154010,000 Tubes
N80105401,000 tubes
Catalog number N8011540
Price (USD)
1,776.00
Each
Quantity:
10,000 Tubes
Price (USD)
1,776.00
Each
The Applied Biosystems™ MicroAmp™ Reaction Tubes with Caps are optimally designed for precise PCR and are designed for use with Applied Biosystems 0.2 mL thermal cyclers including the Veriti™ 96-Well Thermal Cycler, GeneAmp™ PCR System 9700 and the Applied Biosystems 2720 Thermal Cycler.

Features:

• Consistent plastic thickness enables uniform heating and cooling for accurate results.
• Captive lid with positive-click closure assures proper seating.
• Polished surface and conical bottom allows maximum sample recovery.
• Available in economical bulk packages.
• Option of autoclaved tubes provide for a clean, controlled start

Optimized Design
MicroAmp™ Reaction Tubes provide unmatched temperature accuracy and uniformity for fast, oil-free PCR amplification with our 0.2mL 96-Well thermal cyclers. Like all of our products, MicroAmp™ Reaction Tubes undergo stringent quality testing to ensure reproducible results. They ensure the greatest temperature and amplification uniformity for your samples.

Convenient
MicroAmp™ Reaction Tubes with Caps are available as clear or colored and in bulk packs. Use MicroAmp™ Trays for easy sample handling and optimized results on our 0.2 mL 96-well thermal cyclers.
Specifications
Certifications/ComplianceCertified DNA/Rnase Free
For Use With (Equipment)Thermal Cyclers
Format10,000 Tubes
No. of Wells96
Product LineMicroAmp™ 
Quantity10,000 Tubes
Reaction SpeedFast
Volume (Metric) Working0.2 mL
Closure TypeDomed Cap
ColorNatural
MaterialPolypropylene
Product TypeReaction Tube
Volume (Metric)0.25 mL
Unit SizeEach
Contents & Storage
Contains 10,000 MicroAmp™ Reaction Tubes with Caps, 0.2 ml (10 packs of SKU# N8010540, Qty 1,000 per pack). Store at room temperature.