GeneAmp® PCR System 9700, 96-Well Silver - FAQs
GeneAmp® PCR System 9700, 96-Well Silver - FAQs
View additional product information for GeneAmp® PCR System 9700, 96-Well Silver - FAQs (N8050001)
9 product FAQs found
Why are my PCR tubes crushed/deformed after my PCR run?
The heated thermal cycler lid is designed to make sure that optimized pressure is applied on the microplate for an efficient PCR reaction. When using PCR tubes, excess pressure on the tubes, from the heated lid can cause deformation of the tubes. To avoid this, we recommend using the Tray/Retainer set, as shown below:
Cat. No. 4381850, MicroAmp 96-Well Tray/Retainer Set for Veriti Systems – This Tray is for use with MicroAmp Strip Tubes (or MicroAmp Reaction Tubes without Caps, 0.2 mL), with the Veriti Thermal Cycler, SimpliAmp Thermal Cycler, and Proflex PCR System.
Cat. No. 403081, MicroAmp 96-Well Tray/Retainer Set – This Tray is for use with MicroAmp Strip Tubes (or MicroAmp Reaction Tubes without Caps, 0.2 mL), with the 7000 System, 2720 Thermal Cycler, and GeneAmp PCR System 9700.
Cat. No. 4379983, MicroAmp 96-Well Tray for VeriFlex Blocks – This Tray is for use with MicroAmp Strip Tubes (or MicroAmp Reaction Tubes with Caps, 0.1 mL), with the Veriti Thermal Cycler, Veriti Fast Thermal Cycler, SimpliAmp Thermal Cycler, and Proflex PCR System.
Please take a look at this video: How to Use Adapter Tray Retainers in Thermal Cycler Starter Kits (https://www.youtube.com/watch?v=1uh2ZxZJtgs).
Find additional tips, troubleshooting help, and resources within our PCR Thermal Cyclers Support Center.
How do I replace lost calibration files on the GeneAmp PCR System 9700?
A guide to replacing lost calibration files for the GeneAmp PCR System 9700 can be found here: http://tools.thermofisher.com/content/sfs/brochures/cms_042718.pdf.
How do I perform the temperature verification on the GeneAmp PCR System 9700?
Please refer to the guidelines in this protocol(https://tools.thermofisher.com/content/sfs/manuals/cms_070449.pdf) specific to the GeneAmp PCR System 9700 and block type.
How do I program an infinite hold in the GeneAmp PCR System 9700?
To add an infinite hold, set the time for the hold to ‘99.59'. The screen should then default to the infinity symbol for your hold.
Is the GeneAmp PCR System 9700 capable of running a gradient?
No, the GeneAmp PCR System 9700 does not have a gradient feature. Our newer thermal cyclers including the ProFlex PCR System and Veriti Thermal Cycler have better-than-gradient VeriFlex Blocks.
What is the Auto Delta (AutoX) feature on the GeneAmp PCR System 9700?
The AutoDelta feature allows you to incrementally increase or decrease the temperature or hold time with each cycle for a given step within a cycling stage. One common application for this is touchdown PCR.
What is the max block ramp rate of the GeneAmp PCR System 9700?
Please see the max block ramp rates in degrees/C below:
- 96-well aluminum: 2.3
- 96-well gold: 2.6
- 96-well silver: 2.6
- dual 96-well: 1.6
- dual 384-well: 1.8
- 60-well: 1.9
What types of plates or tubes can I use with the GeneAmp PCR System 9700?
The plates and plastics will be specific to the block type. Please refer to the tables here (https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics.html) for a list of compatible plastics, based on your block type.
What types of blocks are available on the GeneAmp PCR System 9700?
The GeneAmp PCR System 9700 offers interchangeable sample blocks in a variety of well formats and materials to accommodate a wide range of applications.
96-Well Aluminum
96-Well Gold
96-Well Silver
Dual 96-Well
Dual 384-Well
60-Well