Does rTth DNA polymerase (Cat. No. N808-0098) have RNase H activity?
No, rTth DNA polymerase does not have RNase H activity.
Search
View additional product information for rTth DNA Polymerase, Enzyme + Buffer Pack - FAQs (N8080098)
2 product FAQs found