RNase Inhibitor - FAQs
RNase Inhibitor - FAQs
View additional product information for RNase Inhibitor - FAQs (N8080119)
1 product FAQs found
Can I order the RNase Inhibitor from the High-Capacity cDNA Reverse Transcription Kit separately?
Yes. You can order the RNase Inhibitor separately as Cat. No. N8080119.