rTth DNA Polymerase and EZ Buffer Pack - FAQs
rTth DNA Polymerase and EZ Buffer Pack - FAQs
View additional product information for rTth DNA Polymerase and EZ Buffer Pack - FAQs (N8080178)
2 product FAQs found
Does rTth DNA polymerase (Cat. No. N808-0098) have RNase H activity?
No, rTth DNA polymerase does not have RNase H activity.
Does rTth DNA polymerase have exonuclease activity?
rTth DNA polymerase contains to 5' to 3' exonuclease but not 3' to 5' exonuclease activity, which means that it has no proofreading activity.