Does Tth DNA Polymerase leave 3' A overhangs on PCR products?
Yes. The products of Tth DNA Polymerase leave 3' A overhangs and therefore are able to be cloned into the TA vectors offered by Invitrogen.
Search
View additional product information for rTth DNA Polymerase, XL and XL Buffer II Pack - FAQs (N8080187)
1 product FAQs found