Does Tth DNA Polymerase leave 3' A overhangs on PCR products?
Yes. The products of Tth DNA Polymerase leave 3' A overhangs and therefore are able to be cloned into the TA vectors offered by Invitrogen.
Search
View additional product information for rTth DNA Polymerase, XL and XL Buffer II Pack (Six Paq) - FAQs (N8080188)
1 product FAQs found