Product Image
Invitrogen™
Large Cellophane
The Large Cellophane is one of the components for the Large Gel Drying Kit for drying PAGE gels. The Large자세히 알아보기
Have Questions?
카탈로그 번호수량
NC2200100/ pkg.
카탈로그 번호 NC2200
제품 가격(KRW)
226,000
온라인 행사
Ends: 30-Sep-2026
258,000
할인액 32,000 (12%)
Each
수량:
100/ pkg.
제품 가격(KRW)
226,000
온라인 행사
Ends: 30-Sep-2026
258,000
할인액 32,000 (12%)
Each
The Large Cellophane is one of the components for the Large Gel Drying Kit for drying PAGE gels. The Large Gel Drying Kit provides a fast and easy way to consistently dry gels without cracking and can accommedate up to 1-4 mini-gel(s) or a large format gel.
For Research Use Only. Not for use in diagnostic procedures.
사양
수량100/ pkg.
배송 조건Room Temperature
제품라인Novex
제품 유형Large Cellophane
Unit SizeEach
구성 및 보관
100/pack; Store at room temperature
자주 묻는 질문(FAQ)
What are the dimensions of the Large Cellophane (Cat. No. NC2200)?
The Large Cellophane (Cat. No. NC2200) is a component of the Large Gel Drying Kit (Cat. No. NI2207). The Cellophane Dimensions are 22.35 cm × 22.85 cm. You will find these dimensions in the manual of the Drying Kit on page 4 (see link: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/geldrying_man.pdf).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
I dried my gel using your DryEase Mini Gel Drying system and the gel cracked during drying. Can you please help?
Please see below for possible causes and solutions:
- Air trapped between cellophane layers. Apply adequate Gel-Drying solution to prevent bubbles between the gel and cellophane assembly.
- Tension on cellophane too strong. Take care not to stretch the cellophane during dryer assembly.
- Rough edges or small cracks on gel edges. Trim rough edges from the gel to eliminate starting points for cracks.
- Cellophane slips during drying. Make sure clamps hold all four edges of the dryer assembly firmly.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
I am using your DryEase Mini Gel Drying system and would like to peel off the cellophane after drying the gel but it seems really difficult. Can you please offer some tips?
Usually it is not necessary to peel off the cellophane. If your signal is very faint and you are concerned about getting an exposure, you can rub the top surface of your gel with a damp towel and then peel off the cellophane. Then air dry the gel for a few minutes and perform autoradiography.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
I dried my gel using Gel-Drying Solution and it dried milky white. Can the gel be recovered?
Gels turn white when they are dried too rapidly, usually in an environment that is too ventilated, drafty, or warm. You can remedy this problem by dabbing the gel with deionized water on a Kimwipe tissue to rehydrate the white areas. Re-dry the gel in a dark, dry, non-drafty place, such as a drawer or cabinet.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What is the pore size of the DryEase cellophane?
The pore size of DryEase cellophane is 40 microns.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.