Empty Gel Cassette Combs, mini, 1.5 mm, 2D well - FAQs
Empty Gel Cassette Combs, mini, 1.5 mm, 2D well - FAQs
View additional product information for Empty Gel Cassette Combs, mini, 1.5 mm, 2D well - FAQs (NC3502)
1 product FAQs found
Are the Invitrogen empty gel cassettes and combs RNase-free?
No. The cassettes and combs are not RNase free. There is no sterility QC test for RNase activity. It may be possible to treat with RNaseZAP, or another RNase decontamination solutions followed by a thorough rinse with RNase-free water.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.