NanoDrop Ultra Security and IQ/OQ Bundles
NanoDrop Ultra Security and IQ/OQ Bundles
NanoDrop Ultra Security and IQ/OQ Bundles
Thermo Scientific™
NanoDrop Ultra Security and IQ/OQ Bundles
Purchase a Thermo Scientific™NanoDrop™Ultra Acclaro Pro Security and IQ/OQ Bundle to receive a NanoDrop Ultra instrument, a model-specific IQ/OQ kit, and SciVault 2 software enabling 21 CFR Part 11 compliance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesFor Use With (Equipment)
ND-ULTRA-SEC-GLNanoDrop Ultra instrument, SciVault 2 software on a USB stick, and IQ/OQ kit.NanoDrop Ultra Spectrophotometers and Fluorometers
ND-ULTRAC-SEC-GLNanoDrop Ultra instrument, SciVault 2 software on a USB stick, and IQ/OQ kit.NanoDrop Ultra Spectrophotometers and Fluorometers
ND-ULTRAFL-SEC-GLUSB flash drive containing the Acclaro Pro software for use with the NanoDrop Ultra Spectrophotometers and Fluorometers NanoDrop FLNanoDrop Ultra Spectrophotometers and Fluorometers
ND-ULTRACFL-SEC-GLUSB flash drive containing the Acclaro Pro software for use with the NanoDrop Ultra Spectrophotometers and Fluorometers NanoDrop UltraC FLNanoDrop Ultra Spectrophotometers and Fluorometers
Catalog number ND-ULTRA-SEC-GL
Price (USD)
-
Includes:
NanoDrop Ultra instrument, SciVault 2 software on a USB stick, and IQ/OQ kit.
For Use With (Equipment):
NanoDrop Ultra Spectrophotometers and Fluorometers
The Thermo Scientific™NanoDrop™Ultra Spectrophotometers and Fluorometers represent the next generation of microvolume detection. This cutting-edge instrument delivers exceptional accuracy and sensitivity for nucleic acid and protein analysis with sample volumes as low as 1.0 μL. The NanoDrop Ultra is designed to operate seamlessly within workflows across a wide range of fields, including genomics, proteomics, molecular biology, biotechnology, pharmaceutical, and life sciences research. It provides unparalleled precision, efficiency, and reliability in one compact device. These bundles include of one of the NanoDrop Ultra instruments, SciVault 2 activation, and a model-specific IQ/OQ kit.
Avoid costly setbacks in your workflow with an understanding of sample quality like never before using the Thermo Scientific™ NanoDrop™ Ultra Microvolume UV-Vis Spectrophotometers and Fluorometers. Innovative Thermo Scientific™ Acclaro™ Sample Intelligence technology is built into every instrument for improved measurement accuracy and contaminant identification. Quantify and qualify DNA, RNA, and protein samples in seconds with only 1-2 μL, and obtain full-spectral data before you decide to use samples in downstream applications. A cuvette position is available for additional flexibility.
Tools to aid in 21 CFR Part 11 compliance
Optional Thermo Scientific™ SciVault™ 2 software installs on either the instrument or a companion PC and gives labs the tools they need to comply with US FDA 21 CFR Part 11.
  • Compliance made easy – integrates into the NanoDrop Ultra software user interface and controls user account management, user account access, digital signatures, and audit logs.
  • On-board options – available to be run directly on the local instrument through on-board user account management or connect your instrument to your domain to use your Windows account management.
  • Centralized solution - install SciVault 2 software on a central computer to control privileges and view audit logs across multiple NanoDrop Ultra instruments in different labs.
Streamlined workflow
  • Fully integrated qPCR recipe calculator - use the built-in library of kits, or add your favorite, to let the software run the math for you. The qPCR recipe calculator suggests how to prepare the reaction based on your sample's concentration.
  • Auto-Measure and Auto-Blank functions – multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF with a single tap on the screen.
Advanced connectivity for data storage, printing and sharing
  • Export to the cloud – export data directly to popular cloud storage services like Microsoft OneDrive, Google Drive, and Thermo Fisher Connect.
  • Direct LIMS integration – send data directly into your LIMS with the assistance of an integrated RESTful API.
  • Easy data-to-printer options – Print your results directly from the NanoDrop Ultra instrument via Wi-Fi, Ethernet or USB printer connection; print exported data from your PC using the Nanodrop Ultra PC Control software.
  • Data Your Way – customize and preview the information included in the data file prior to export to ensure it meets any downstream needs.
Model-specific IQ/QQ Kits
Includes:
  • USB flash drive containing IQ/OQ procedure for NanoDrop Ultra
  • CHEM-PR1-KIT
  • CHEM-PV-1
  • Cuvette Qualification Kit (840-257300) if cuvette model purchased.
  • CHEM-FL-1 if fluoresence model purchased.
For research use only; not for diagnostic purposes.
Specifications
For Use With (Equipment)NanoDrop Ultra Spectrophotometers and Fluorometers
IncludesNanoDrop Ultra instrument, SciVault 2 software on a USB stick, and IQ/OQ kit.
Product LineNanoDrop
TypeND Ultra and Security Package
Unit SizeEach