2000c packaged with laptop computer - FAQs
2000c packaged with laptop computer - FAQs
View additional product information for NanoDrop™ 2000/2000c Spectrophotometers - FAQs (ND-2000, ND2000LAPTOP, ND-2000C, ND2000CLAPTOP)
42 product FAQs found
How do I check the accuracy of my NanoDrop 2000/2000c Spectrophotometer?
Our NanoDrop CF-1 Calibration Fluid should be used with the Calibration Check module of the instrument software. CF-1 is prepared from the NIST potassium dichromate standard SRM935 in acidified reagent grade water (Certificate of Analysis available from NanoDrop products).
What additional analytes or biomolecules are routinely analyzed using the NanoDrop 2000/2000c Spectrophotometer?
Our NanoDrop 2000/2000c Spectrophotometers are routinely used to determine the concentrations of dsDNA, RNA, ssDNA, fluorescent dyes incorporated into microarray and proteomic probes, microbial cell cultures and virtually any analyte that exhibits absorbance in the 190-840 nm range.
Can I simply wipe the pedestal surface enough to prevent carryover on my NanoDrop 2000/2000c Spectrophotometer?
Yes, we recommend using a dry laboratory wipe as a very effective method for removing the sample. The highly polished quartz and stainless steel surfaces of the sample retention system are resistant to sample adherence.
What level of reproducibility should I expect from the NanoDrop 2000/2000c spectrophotometers?
The following is an example of the level of reproducibility that can be expected from the NanoDrop 2000/2000c Spectrophotometers:
BSA in A280 module:
- +/- 0.1 mg/mL for samples < 10 mg/mL
- +/- 2% for samples > 10 mg/mL
What sort of accuracy should I expect with the NanoDrop 2000/2000c spectrophotometers?
Typically, the accuracy of NanoDrop 2000/2000c Spectrophotometers falls within 2%.
What is the dynamic (concentration) range for proteins that can be measured on my NanoDrop 2000/2000c spectrophotometers?
BSA concentrations between 0.1 mg/mL to 400 mg/mL can be measured using the A280 method on the NanoDrop 2000/2000c Spectrophotometers.
Refer to the user manual or our website for additional information regarding the detection limits for the Proteins and Labels module as well as for the various colorimetric assays:
https://assets.thermofisher.com/TFS-Assets/CAD/manuals/NanoDrop-2000-User-Manual-EN.pdf
What sample size should I use for the NanoDrop 2000/2000c spectrophotometers?
We recommend using a 2 µL sample size for pedestal-based protein measurements with NanoDrop 2000/2000c Spectrophotometers. Proteins and/or protein buffers may alter the surface tension properties of the solution and using the larger sample size is recommended to ensure proper column formation.
As little as 0.5 µL can be used for precious high concentration samples.
I am using a colorimetric method (e.g. Bradford, BCA etc.) to determine my protein concentration. Can I measure my sample using the A280 method on the NanoDrop 2000/2000c spectrophotometers?
The A280 method is most applicable to purified proteins. Colorimetric assays such as BCA, Pierce 660 nm, Bradford, and Lowry are generally used for uncharacterized protein solutions and cell lysates.
If you are using a colorimetric assay now it is recommended that you continue to do so, and then run the sample (using the appropriate application) on the NanoDrop 2000/2000c to read absorption.
If you are using a Bradford assay, you may find that the Pierce 660 nm assay is a better alternative.
Can I quantify proteins using the NanoDrop 2000/2000c Spectrophotometers?
Yes, you can quantify proteins using the NanoDrop 2000/2000c Spectrophotometers.
Available methods/modules include:
- A280 method for purified proteins
- Proteins and Labels module for labeled antibodies and other protein incorporating fluorescent labels or intrinsic fluorescence
- Colorimetric assays such as the BCA, Bradford, Modified Lowry and Pierce Protein 660 nm assay
- Custom methods may also be set up via the Method Editor to analyze proteins, including peptides at 205 nm
- Proteins may be quantified using a micro-volume sample on the pedestal (2000 and 2000c) or using a standard cuvette (2000c).
What is the Beer-Lambert equation?
Beer-Lambert Equation is as follows:
A = E * b * c
- A is the absorbance of the sample
- E is the wavelength-dependent molar absorptivity coefficient (or extinction coefficient) expressed in units of L/mol-cm
- b is the pathlength in cm
- c is the analyte concentration in mol/L or molarity (M).
For Nucleic Acid calculations, the Beer-Lambert equation is modified to use an extinction coefficient with units of ng-cm/mL. Using this extinction coefficient gives a manipulated equation:
c = (A * e)/b
- c is the nucleic acid concentration in ng/µL
- A is the absorbance of the sample
- e is the nucleic acid specific concentration factor in ng-cm/µL (dsDNA = 50 ng/µL; ssDNA = 33 ng/µL; RNA = 40 ng/µL; dsRNA = 40 ng/µL)
- b is the pathlength in cm
When using a NanoDrop spectrophotometer, what does 10 mm equivalent absorbance mean?
NanoDrop spectrophotometers use light pathlengths from 1.0 mm to as low as 0.05 mm (model dependent) to make measurements, allowing the quantification of samples up to 50 or 200 times more concentrated than traditional cuvette based spectrophotometers are able to measure. Applications such as Nucleic Acid and Protein A280 will display this absorbance as a 10 mm equivalent absorbance.
Are the sample concentrations I obtain with the NanoDrop spectrophotometer pedestal volume dependent?
The sample concentrations are calculated according to Beer's Law which is volume independent. Beer's Law relates absorbance to concentration using a wavelength specific molar extinction coefficient or the use of a nucleic acid concentration factor and a pathlength of 1 cm.
Which pathlength was used for my sample measurement on the NanoDrop spectrophotometer pedestal?
The software is programmed to determine the best pathlength for the measurement based upon the absorbance signal of the sample at the analysis wavelength for the Nucleic Acids module, Protein A280 module, and Custom methods. On the other hand, the autopathlength function in the UV-Vis module monitors the entire wavelength range to optimize pathlength selection. The absorbance signal is normalized to a either 10 mm or 1.0 mm pathlength equivalent depending on module.
When using a NanoDrop spectrophotometer, how does data collected in UV-Vis module compare to the data collected in another application?
UV-Vis data is reported at a normalized 1.0 mm pathlength. As a result, absorbance values reported here will be reported 10-fold lower than those reported by the Nucleic Acid or Protein A280 applications.
When using a NanoDrop spectrophotometer, when do I use the autopathlength function in the UV-Vis module?
Selecting autopathlength should be considered if the peak of interest is greater than 1.25 A at a 1.0 mm pathlength or to ensure that the entire sample absorbance spectrum remains within the linear response of the detector. With autopathlength deselected, the instrument uses only a 1.0 mm pathlength which is optimized for absorbance signal less than 1.25 A.
When using a NanoDrop spectrophotometer, what are the advantages of the Pierce 660 assay over the Bradford assay?
Both assays are mix-and-measure (minimal incubation times and no heating steps). Pierce 660 Assay has a more stable end-point. Pierce 660 reaction will not continue to progress and form aggregates.
When performing a Pierce 660 assay with a NanoDrop spectrophotometer, why am I experiencing lower absorbance on the NanoDrop pedestal compared to the cuvette?
When utilizing the measurement pedestal, NanoDrop spectrophotometers will report the Pierce 660 assay absorbance using a 1.0 mm pathlength. As a result, the absorbance displayed will be 10-fold lower than if measured in a conventional 10 mm cuvette. This reduction in signal does not compromise the sensitivity or linear range of the assay.
When performing a Lowry protein assay, why am I experiencing low absorbance signal across the dynamic range of the assay when using a NanoDrop spectrophotometer?
The Lowry module reports absorbance signal at a 1.0 mm pathlength rather than the conventional 10 mm pathlength resulting in a 10-fold reduction in absorbance signal. Consequently, the reduction in signal will not compromise the sensitivity or dynamic range of the assay. In the cases of extreme low signal, it is typically due to the formulation of the Lowry reagent from the specific vendors. We have had the greatest success with the Pierce product line of colorimetric assays.
General Information: Wavelength (modified Lowry protocol for a NanoDrop spectrophotometer) is measured at 650 nm and normalized at 405 nm Dynamic range of assay is 0.2 mg/mL to 4.0 mg/mL (BSA)
How can insufficient mixing affect my Bradford assay when using a NanoDrop spectrophotometer?
A poorly mixed Bradford assay will not properly complete and may form a gradient in the tube. This can affect reproducibility.
Why am I experiencing low absorbance formulations, when performing Bradford assays with my NanoDrop spectrophotometer?
Some Bradford assay manufacturers' formulations will not provide sufficient absorbance to obtain an accurate and reproducible standard curve.
Why is the reproducibility of my Bradford assay outside the manufacturer's specification with a NanoDrop spectrophotometer?
The Bradford reagent will form particulates as a reaction progresses. These particulates may cause light scatter during a sample measurement, resulting in higher than expected variation between measurements. Particulates can be minimized by limiting the incubation time to 10 min prior to measuring as per the Thermo Scientific Pierce protocol. In addition, a sample volume of 2 µL should be used for all colorimetric assay measurements.
What is the importance of uniform sample heating when performing a BCA assay with a NanoDrop spectrophotometer?
To ensure accurate results, sample heating must be uniform for both standards and unknown samples.
When performing a BCA assay with a NanoDrop spectrophotometer, why am I obtaining a lower absorbance signal on the NanoDrop spectrophotometer pedestal compared to a conventional cuvette system?
When utilizing the measurement pedestal, NanoDrop spectrophotometers will report the BCA assay absorbance using a 1.0 mm pathlength. As a result, the absorbance displayed will be 10-fold lower than if measured in a conventional 10 mm cuvette. This reduction in signal does not compromise the sensitivity or linear range of the assay.
When I am using a NanoDrop spectrophotometer, what is the appropriate ratio for a pure protein sample?
A purified protein should show a 260/280 purity ratio of approximately 0.63.
Why is it not recommended that I use RIPA buffer for protein A280 measurements with my NanoDrop spectrophotometer?
RIPA buffer produces a particularly strong absorbance signal at the 280 nm wavelength. As a result, it will either over estimate or under estimate protein concentrations and interfere with the protein purity ratio.
Protein samples in RIPA buffer should be quantified via the Pierce Protein 660 or BCA colorimetric assays using a full spectrum NanoDrop model.
Find additional tips, troubleshooting help, and resources within ourProtein Purification and Isolation Support Center.
When I am using a NanoDrop spectrophotometer to calculate protein concentrations, what is the difference between E1% and E0.1% value?
The difference between the E1% and E0.1% values are the associated units for the results.
The E1% value has units of a 1% solution or 1 g/100mL. In the NanoDrop software this value is converted to an E0.1% value to display protein concentrations in mg/mL units.
Please refer to Tech Tip #6: Extinction Coefficients for further detail: https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0006-Extinction-coefficients.pdf
How are protein concentrations calculated by the Protein A280 module in the NanoDrop spectrophotometer?
Protein concentrations obtained from the Protein A280 module are calculated using a 1% solution extinction coefficient (E1%) in place of the molar extinction coefficient. Each purified protein will have a unique E1% value which can be obtained from either references or, if produced commercially, from the vendor. The user may choose to use a known E1% value or input the molar extinction coefficient and molecular weight of the protein into the NanoDrop software to obtain the mg/mL protein concentrations.
If only a rough estimation is desired, the 1 Abs = 1 mg/mL can be used instead of a specific E1% value.
How do I perform a Cuvette Calibration Check for my NanoDrop 2000/2000c Spectrophotometer?
Although a specific cuvette calibration check procedure is not available for the NanoDrop 2000c, we suggest the use of a 10%T and 50%T neutral density filter.
When performing a calibration check on my NanoDrop 2000/2000c Spectrophotometer, what does "conditionally passing" mean?
A conditional pass indicates that the calibration check % error for the 1.0 mm pathlength was >3% but = 5%.
If recalibration is required contact technical support to arrange for service.
My NanoDrop 2000/2000c Spectrophotometer failed the calibration check. What should I do?
Please review the following steps to take when/if your NanoDrop 2000/2000c fails a calibration check:
- Clean the pedestal with the NanoDrop PR-1 Reconditioning Kit
- Repeat calibration check using a FRESH vial of NanoDrop CF-1 Calibration Fluid and FRESH blanking solution
More detailed instructions can be found on the Calibration Check Procedure document: https://assets.thermofisher.com/TFS-Assets/CAD/Product-Bulletins/NanoDrop-2000-2000c-1000-Calibration-Check-procedure.pdf
If your NanoDrop 2000/2000c continues to be out of specification, Contact Technical Support.
Where do I find the Calibration Check Software for my NanoDrop 2000/2000c Spectrophotometer?
The Calibration Check software for the NanoDrop 2000/2000c is included in the operating software. Select the Diagnostics stacked task bar from the software home page and then select the Calibration Check stacked task bar from the expanded menu.
Will someone come on-site to calibrate my NanoDrop 2000/2000c Spectrophotometer or do I need to send it in?
All US & Canada instrument calibrations are performed in our Madison, Wisconsin service center by trained NanoDrop service technicians. There is a 48 hr turnaround time for all domestic service. International customers should contact their local distributors for service. The NanoDrop 2000/2000c must be shipped in the original box and packaging. Please contact Technical Support or your local distributor if you require a box for shipment.
How often should I check the calibration of my NanoDrop 2000/2000c Spectrophotometer?
We recommend you check the calibration of your NanoDrop 2000/2000c every 6 months using the NanoDrop CF-1 Calibration Fluid.
How do I check the calibration of my NanoDrop 2000/2000c Spectrophotometer? Do I need to calibrate it?
You can easily check the calibration of your NanoDrop 2000/2000c Spectrophotometer by using NanoDrop CF-1 Calibration Fluid. CF-1 is specifically designed for NanoDrop spectrophotometers and is available from the Thermo Fisher website (Cat. No. CHEM-CF-1).
If re-calibration is ever needed, it is performed by a trained service technician. Due to the precise nature of the NanoDrop instrument design, these spectrophotometers seldom require re-calibration.
Do I need to recondition the pedestals of my NanoDrop spectrophotometer?
Pedestal surfaces may become "un-conditioned" and possibly inhibit proper liquid column formation required for measurement precision. Refer to this Cleaning and Reconditioning document for more information: http://tools.thermofisher.com/content/sfs/brochures/T005-NanoDrop%201000-&-NanoDrop%208000-Cleaning-and-Reconditioning.pdf
What if I use my NanoDrop spectrometer to measure a sample that is too diluted?
Measuring at or below the detection limit may result in SD values outside the precision specification. Refer to instrument user manual for detection limits for specific applications.
Can I make multiple measurements on a single aliquot when using a NanoDrop spectrophotometer?
Evaporation can occur when samples remain on the pedestal beyond the time required to make one measurement, therefore we recommend always using a fresh aliquot for each measurement.
Why am I experiencing poor sample homogeneity when using my NanoDrop spectrophotometer?
Poor sample homogeneity is commonly associated with non-homogeneous DNA samples. Due to the small sample volumes used for pedestal measurements it is important to mix the sample thoroughly. Samples containing large molecules such as genomic or lambda DNA are particularly susceptible to this phenomenon. If compatible with the protocol being used, heat the DNA samples to 63 degrees C and lightly finger vortex before measurement to ensure the nucleic acid is properly in solution.
When using a NanoDrop spectrophotometer, what is the difference between controls versus standards?
NanoDrop CF-1 Calibration Fluid is the only photometric standard available to assess the accuracy of the NanoDrop spectrophotometer pedestal. If the control sample shows unexpected results the user should confirm the calibration of the instrument with the CF-1 fluid.
Some control examples include:
- Nucleic Acids: Fermentas GeneRuler Express DNA Ladder 100-5000 bp (Cat. No. SM1551)
- Protein: Pierce Bovine Serum Albumin Standard Ampules, 2 mg/mL (Cat. No. 23209)
How can I check the accuracy of my NanoDrop spectrophotometer?
The accuracy of NanoDrop spectrophotometers can be assessed by using NanoDrop CF-1 Calibration Fluid in conjunction with the calibration check application in the NanoDrop software.
What are acceptable purity ratios, when using a NanoDrop spectrophotometer?
Please review the following information regarding acceptable purity ratios, when using NanoDrop spectrophotometers:
- Nucleic acid samples: A260/A280 ratios of 1.8-2.2; A260/A230 ratios of 1.8-2.2 are generally considered pure
- Protein samples: A260/A280 ratio of approximately 0.63 is generally considered pure
When using a NanoDrop spectrophotometer, what would cause my 260/280 purity ratios to fall outside of the generally accepted range?
Please review the following possible causes for 260/280 purity ratios to fall outside of the generally accepted range when using a NanoDrop spectrophotometer:
- The starting sample or the extraction method may result in contaminants in the final extract, e.g. guanidine (often from column based kits), phenol carryover or carbohydrates from the original cells (often a problem with plant samples). This is often best diagnosed by looking at the spectra, as many contaminants have characteristic profiles.
- Improper Blank: An incorrect or highly absorbent blanking solution or sample residue on the measurement pedestal may affect purity ratios.
- Sample Concentration: If the sample is nearing the lower detection limit of the instrument. Contributions from instrument noise may affect the shape of the spectrum. Check instrument specifications for lower detection limits. Purity ratios cannot be accurately calculated for samples close to the lower detection limits of the instrument. If a NanoDrop 2000c is being used, a cuvette may be used for samples at very low concentrations.