NanoDrop Lite Plus Spectrophotometers
Thermo Scientific™

NanoDrop Lite Plus Spectrophotometers

Quickly and easily assess sample concentration and purity ratios with the entry-level Thermo Scientific™ NanoDrop™ Lite Plus Microvolume Spectrophotometer. The instrument delivers accurate measurements in an easy-to-use, affordable package. Ideal for teaching and busy research labs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesPrinter Included
NDLPLUSGLND Lite PlusNo
NDLPLUSPRGLND Lite PlusYes
Catalog number NDLPLUSGL
Price (KRW)
-
Includes:
ND Lite Plus
Printer Included:
No

Quickly and easily assess sample concentration and purity ratios with the Thermo Scientific™ NanoDrop™ Lite Plus Microvolume Spectrophotometer, the entry-level complement to the full-featured Thermo Scientific™ NanoDrop™ One/OneC and NanoDrop™ Eight Spectrophotometers. While offering fewer features than our premium models, this instrument delivers fast, accurate measurements in an easy-to-use, affordable package.

The NanoDrop Lite Plus Spectrophotometer is a valuable teaching tool and is ideal for quality check measurements in busy research labs as well. Measure purified DNA, RNA, and protein concentration up to 30 Abs and calculate critical A260/A280 and A260/A230 purity ratios using trusted NanoDrop technology.

The NanoDrop Lite Plus Spectrophotometer is designed for routine measurements of nucleic acid and protein samples using only 1–2 μL. Our pioneering sample-retention technology eliminates the need to dilute highly concentrated samples, making measurements easy. Simply pipette your sample directly onto the optical measurement pedestal, measure, and wipe clean with a lint-free wipe. Its compact design, with built-in controls and software, means the NanoDrop Lite Plus Spectrophotometer takes up little space on your benchtop.

With an intuitive user interface and modern industrial design, the NanoDrop Lite Plus Spectrophotometer is the instrument of choice for basic microvolume measurements. Even if budgets are tight, your lab can have a NanoDrop, too.

Confidence in your results

  • Standard measurements
    • Measures purified DNA, RNA, and protein concentration up to 30 Abs
      • Up to 1,500 ng/μL dsDNA
      • Up to 1,200 ng/μL RNA
      • Up to 45 mg/mL BS
    • Calculates critical A260/A280 and A260/A230 purity ratios
  • Fast, accurate measurements from only 1–2 μL—saves precious samples, no waste
    • Sample-retention technology uses surface tension to hold the sample in place—no cuvettes or expensive consumables are needed
  • Intuitive interface
    • Built-in touchscreen operation
    • No computer required
  • Data storage and transfer
    • Last 1,000 measurements saved locally; export data to a USB device
  • Optional printer available for freezer-compatible labels (not included)
  • Compact and portable—fits on any lab bench, ready for your next experiment
  • Affordably priced—stretch tight budgets

Reach your goals

  • Academic teaching—educate students on molecular biology principles and experimental techniques on an industry-leading instrument
  • Academic research labs—facilitate your research and publication of your novel research findings
  • Biotech start-ups—accelerate the develop of new products
Specifications
Absorbance Accuracy3% at 0.97 A, 302 nm, 23 ± 2°C Abs/mm at 25 °C
Absorbance Range0.04 to 30 Abs (10 mm equivalent)
Certifications/ComplianceUL/CSA and CE
CompatibilityLocal Control Only
Concentration1,500 ng/μL (1,200 ng/μL) dsDNA (RNA)45 mg/mL (21 mg/mL) BSA (lgG)
ConnectionsUSB 2.0 Port
Depth (Metric)22 cm
Detection Limits2.0 ng/μL (1.6 ng/μL) dsDNA (RNA)0.06 mg/mL (0.03 mg/mL) BSA (lgG)
Footprint27 x 22 x 22 cm (0.6 x 8.7 x 8.7 in); W x D X H
For Use With (Application)Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification
Height (Metric)22 cm
IncludesND Lite Plus
MaterialStainless Steel (303), Quartz Fiber
Measurement Time≤ 5 seconds
No. of Samples1
Pathlength1.0 mm and 0.2 mm (auto adjust)
Power Consumption18 W (standby < 3 Watts)
Printer IncludedNo
ProcessorStandalone control with data export via USB Flash Drive
Sample Volume (Metric)1 to 2 μL
Spectral Resolution≤ 1.8 nm (FWHM at Hg 254 nm)
System RequirementsStandalone control with data export via USB Flash Drive
WarrantyTwo year
Wavelength Accuracy± 1 nm
Weight (English)6.0 lbs (without printer)
Weight (Metric)2.7 kg (without printer)
Width (Metric)27 cm
Detector Type2048-element CMOS Linear Image Sensor
DisplayLCD Touchscreen
Light SourceXenon Flash
Product LineMicrovolume
TypeMicrovolume UV-Vis Spectrophotometer
Voltage100/240 V
Wavelength Range230 nm, 260 nm, 280 nm
Unit SizeEach

Frequently asked questions (FAQs)

How do I obtain service for my NanoDrop Lite Plus?

For service in the United States and Canada, please call 877-724-7690 option 3 or email nanodrop@thermofisher.com.
For international service, please contact your local NanoDrop distributor.

Does the NanoDrop Lite Plus come with 21 CFR Part 11 compliance software?

No. It does not align with 21 CFR Part 11. If this is needed, please reach out to us about the NanoDrop Ultra family.

What verification standards should I use with NanoDrop Lite Plus?

Use the NanoDrop PV-1 Performance Verification solution with the instrument’s verification routine (see manual).

How do I clean and maintain the pedestal with NanoDrop Lite Plus?

The NanoDrop Lite Plus user guide has step-by-step cleaning, decontamination, and reconditioning procedures; use lint-free wipes and follow the recommended solvents and safety steps.

What analyses does the NanoDrop Lite Plus provide?

It provides concentration measurements for DNA, RNA, and purified proteins at 230 nm, 260 nm, and 280 nm, plus A260/A280 and A260/A230 purity ratios for QC checks. This instrument does not provide full spectral data, nor does it include Acclaro Sample Intelligence.