How do I obtain service for my NanoDrop Lite Plus?
For service in the United States and Canada, please call 877-724-7690 option 3 or email nanodrop@thermofisher.com.
For international service, please contact your local NanoDrop distributor.
Does the NanoDrop Lite Plus come with 21 CFR Part 11 compliance software?
No. It does not align with 21 CFR Part 11. If this is needed, please reach out to us about the NanoDrop Ultra family.
What verification standards should I use with NanoDrop Lite Plus?
Use the NanoDrop PV-1 Performance Verification solution with the instrument’s verification routine (see manual).
How do I clean and maintain the pedestal with NanoDrop Lite Plus?
The NanoDrop Lite Plus user guide has step-by-step cleaning, decontamination, and reconditioning procedures; use lint-free wipes and follow the recommended solvents and safety steps.
What analyses does the NanoDrop Lite Plus provide?
It provides concentration measurements for DNA, RNA, and purified proteins at 230 nm, 260 nm, and 280 nm, plus A260/A280 and A260/A230 purity ratios for QC checks. This instrument does not provide full spectral data, nor does it include Acclaro Sample Intelligence.
Is a computer required to run the NanoDrop Lite Plus?
No. The NanoDrop Lite Plus is a stand-alone instrument with preloaded software so it can be used without a computer; optional printer accessories are available.
What is the required sample volume for NanoDrop Lite Plus?
The NanoDrop Lite Plus is designed for routine microvolume measurements using 1–2 µL sample volumes.