Search
| Catalog Number | Includes |
|---|---|
| NDUC-Q4 | NanoDrop UltraC Spectrophotometer & Qubit 4 Fluorometer |
| NDU-Q4 | NanoDrop Ultra Spectrophotometer & Qubit 4 Fluorometer |
As a bundled offering, NanoDrop Ultra/UltraC spectrophotometers and the Qubit 4 Fluorometer together allow you to obtain the most complete information about the concentration and quality of your DNA, RNA, or protein samples, helping prevent costly troubleshooting downstream. While both the NanoDrop Ultra Spectrophotometer and the Qubit 4 Fluorometer offer accurate measurements of DNA, RNA, and protein samples, each also has its unique advantages.
NanoDrop Ultra and UltraC spectrophotometers
Thermo Scientific NanoDrop™ Ultra and UltraC spectrophotometers represent the next generation of microvolume detection. These cutting-edge instruments deliver exceptional accuracy and sensitivity for nucleic acid and protein analysis with sample volumes as low as 1.0 μL. The NanoDrop Ultra Spectrophotometer is designed to streamline workflows across a wide range of fields, including genomics, proteomics, molecular biology, biotechnology, pharmaceutical research, and life sciences research. It provides unparalleled precision, efficiency, and reliability in one compact device.
Features of the NanoDrop Ultra/UltraC spectrophotometers include:
Qubit 4 Fluorometer
The easy-to-use Qubit 4 Fluorometer is a benchtop fluorometer that can be used for the quantitation of DNA, RNA, microRNA, and protein, as well as the measurement of RNA integrity and quality using the highly sensitive and accurate fluorescence-based Qubit assays. The fluorometer can also directly measure the fluorescence of samples or help create new assays using the MyQubit software and web tool. The Qubit 4 Fluorometer is ideal for sensitive applications including next-generation sequencing (NGS) and qPCR.
Features of the Qubit 4 Fluorometer include: