Neon™ NxT Electroporation System 1-Channel Tubes, 24 Tubes - FAQs
Neon™ NxT Electroporation System 1-Channel Tubes, 24 Tubes - FAQs
View additional product information for Neon™ NxT Electroporation System 1-Channel Tubes - FAQs (NT96, NT24)
1 product FAQs found
How are the Neon NxT Electroporation System 1-Channel Tubes (Cat. No. NT24 and NT96) packaged?
The tubes are packaged in groups of 4. Therefore, Cat. No. NT24 will have 6 bags of 4 tubes and Cat. No. NT96 will have 24 bags of 4 tubes.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.