MAS™ Omni·IMMUNE™ Immunoassay Controls
Actual product may vary
Thermo Scientific™
MAS™ Omni·IMMUNE™ Immunoassay Controls
Consolidate routine immunoassay, tumor marker and specialty immunoassay QC processing into a single product offering three distinct levels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevel
OIM-3033
OIM-1011
OIM-2022
Catalog number OIM-303
Price (MXN)
-
Level:
3
Consolidate routine immunoassay, tumor marker and specialty immunoassay QC processing into a single product with Thermo Scientific™ MAS™ Omni·IMMUNE Immunoassay Controls.
  • Liquid, ready-to-use
  • Human serum based matrix
  • Value assignment provided for most major analyzer
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionOmni IMMUNE Control, Assayed
Level3
Packaging6 Pk.
Quantity5 mL
Shelf Life36 months at -25° to -15°C
TypeControl
Detectable Analytes17-OH-Progesterone, 25-OH-Vitamin D, Acetaminophen, Adrenocorticotropic Hormone (ACTH), Aldosterone, alpha-Fetoprotein (AFP), Amikacin, Benzodiazepines, beta-2-microglobulin (B2M), β-hCG, CA-125, CA 15-3, CA 19-9, Carbamazepine, Carcinoembryonic Antigen (CEA), Cortisol, C-peptide, DHEA Sulfate, Digoxin, Disopyramide, Estradiol, Estriol (Free), Ethosuximide, Ferritin, Folate (Vitamin B9), Fructosamine, Follicle Stimulating Hormone (FSH), Gastrin, Gentamicin, Growth Hormone, hCG, Homocysteine, IgE, Insulin-like Growth Factor 1 (IGF-1), Insulin, Luteinizing Hormone (LH), Lidocaine, Lithium, N-acetylprocainamide (NAPA), Prostatic Acid Phosphatase (PAP), Phenobarbital, Phenytoin, Phenytoin (Free), Primidone, Procainamide, Procalcitonin (PCT), Progesterone, Prolactin, PSA (Free), PSA (Total), Parathyroid Hormone (PTH), Quinidine, Salicylate (SALY), T3 (Free), Triiodothyroxine (T3 Total), T4 (Free), Thyroxine (T4 Total), Thyroxine-binding Globulin (TBG), Testosterone, Theophylline, Thyroglobulin, Tobramycin, Tricyclic Antidepressants, Thyroid Stimulating Hormone (TSH), T-uptake, Valproic Acid, Valproic Acid (Free), Vancomycin, Vitamin B12
Sample TypeSerum
Unit SizeEach
Contents & Storage
-25°C to -15°C