MAS™ Omni·IMMUNE™ Max Controls
MAS™ Omni·IMMUNE™ Max Controls
Provides consolidation for routine immunoassay, tumor marker and specialty immunoassay QC processing into a single product, offering three distinct levels in a barcoded load and go tube format.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerFüllstand
OIMM-3033
OIMM-1011
OIMM-2022
Katalognummer OIMM-303
Preis (EUR)
-
Füllstand:
3
Consolidate routine immunoassay, tumor marker and specialty immunoassay QC processing into a single product offering three distinct levels with Thermo Scientific MAS Omni·IMMUNE Max Controls. Value assignment is provided for key integrated instrument systems. Load-and-Go barcoded
  • Three distinct levels
  • Liquid, ready-to-use
  • Human-serum based matrix
  • Assayed values available for most analyzers
Available in select countries only
Für In-vitro-Diagnostik
Specifications
EinwegproduktReusable
Füllstand3
SterilitätNon-sterile
TypClinical Chemistry Control
FormLiquid Ready-to-Use
ProbentypHuman Serum
Unit SizeEach