How do I back up my results from BioPharma Finder Software?
All of the results are saved inside this folder C:\ProgramData\ThermoScientific.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.