How do I create and install mass lists for the Search Mass List node in Compound Discoverer?
Please watch this video.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.