Compound Discoverer 3.0 Upgrade from Metworks or Mass Frontier (All Versions) - FAQs
Compound Discoverer 3.0 Upgrade from Metworks or Mass Frontier (All Versions) - FAQs
View additional product information for Compound Discoverer™ Software - FAQs (OPTON-30860)
7 product FAQs found
How do I create and install mass lists for the Search Mass List node in Compound Discoverer?
Please watch this video.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
How do I use the Pattern Tracer and Pattern Scoring Nodes in Compound Discoverer Software?
Please watch this video.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Can you walk me through an Example Study Set in Compound Discoverer Software?
Please watch these videos.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
How do I create a Workflow method to process data in Compound Discoverer Software?
Please watch this video.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
How do I create a new study using the Study Wizard in Compound Discoverer Software?
Please watch this video.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
I am a new user of Compound Discoverer Software. What are the basics I should know?
Please watch this video.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.