Oretronic IV 6 Tramp Metal Detector - FAQs
Oretronic IV 6 Tramp Metal Detector - FAQs
View additional product information for Thermo Scientific™ Ramsey™ Oretronic IV Tramp Metal Detector - FAQs (ORETRONICIV4, ORETRONICIV6)
29 product FAQs found
What regulatory approvals should tramp metal detectors have?
Tramp metal detector approvals should include cCSAus, CE, and RCM.
How fast can tramp metal detectors run?
Oretronic Tramp Metal Detectors (https://www.thermofisher.com/order/catalog/product/ORETRONICIV4) can be used on conveyors with a belt speed of 1.5 m/min to 550 m/min (5 ft/min to 1800 ft/min).
Will tramp metal detectors interfere with other electrical equipment on my conveyor?
Tramp metal detectors should feature variable frequency to eliminate interference from other electrical equipment, and password protection for security.
Can tramp metal detectors withstand harsh conditions?
Most tramp metal detectors are designed for belt conveyors moving coal, iron pellets, minerals, cement, aggregates, and other bulk materials. In the case of Oretronic Tramp Metal Detectors (https://www.thermofisher.com/order/catalog/product/ORETRONICIV4), the system coils are made of impact-resistant polyvinyl chloride and designed to withstand stresses of up to ten times the force of gravity. The transmitter coil and dual-wound receiver coils mount on a support made of non-conductive materials. The transmitter coil is swing-mounted to protect the coil and support from being struck from oversized materials. A dual receiver coil is utilized to counteract interference from external sources, such as electric motors, lightning, and radio transmitters.
The control unit enclosure for the Oretronic IV-4 is made with reinforced fiberglass that conforms to NEMA-4X, IP66 dust, and water tight 432 mm (17 in) x 360 mm (14.2 in) x 184 mm (7.2 in). The Oretronic IV-6 control has painted mild steel that conforms to NEMA-4, IP66 dust, and water tight 687 mm (27 in) x 524 mm (20.6 in) x 230 mm (9.1 in).
Can tramp metal detectors work in wet conditions?
Yes. Many tramp metal detectors, including the Oretronic Tramp Metal Detector (https://www.thermofisher.com/order/catalog/product/ORETRONICIV4), can detect tramp metal when buried in wet conductive materials.
Is there a safety instrument to warn of tramp metal on the conveyor?
Tramp metal detectors protect expensive crushers, conveyors, and other process equipment from damage by tramp metal. The Thermo Scientific Ramsey Oretronic Tramp Metal Detector (https://www.thermofisher.com/order/catalog/product/ORETRONICIV4), for example, is designed for belt conveyors moving coal, iron pellets, minerals, aggregates, and other bulk materials, and can detect all types of metallic scrap that conventional metal detectors may miss, including bucket teeth, manganese steel mantles, bore crowns, bar scrap, chains, and tools.
Why is tramp metal dangerous?
Tramp metal can damage expensive crushers, conveyors, and other process equipment. As a result, tramp metal can stop operations and cause lost production time.
What is tramp metal?
Tramp metal is metallic scrap that may accidentally find its way onto conveyors and into bulk materials. Some examples of tramp metal include bucket teeth, manganese steel mantles, bore crowns, bar scrap, chains, and tools.
Will the Oretronic IV Tramp Metal Detector (TMD) controls be adversely affected if they are mounted in direct sunlight?
We do not recommend mounting the Oretronic IV Tramp Metal Detector (TMD) controls in an area where they will be exposed to high heat or direct sunlight as this may result in darkening of the display with limited visibility or display failure. To avoid exposure to direct sunlight, a sun shield should be installed.
What gauge wire is used with Oretronic IV Tramp Metal Detector (TMD) controls?
Please refer to the user manual (https://assets.thermofisher.com/TFS-Assets/CAD/manuals/tramp-metal-detector-oretronic-iv-s-manual-REC4479B) for all wiring information.
What are the different language options for the Oretronic IV Tramp Metal Detector (TMD) control user manual?
The Oretronic IV Tramp Metal Detector (TMD) control user manual is available in English, Spanish (Latin Am), German, Chinese (Mandarin/Simplified), Italian, and French languages. Please contact the factory at sales.bulk.us@thermofisher.com for other languages.
What are the different language options for the Oretronic IV Tramp Metal Detector (TMD) control software?
The Oretronic IV Tramp Metal Detector (TMD) control software is available in English, Spanish (Latin Am), German, Chinese (Mandarin/Simplified), Italian, and French languages. Please contact the factory at sales.bulk.us@thermofisher.com for other languages.
What approvals do the Oretronic IV Tramp Metal Detector (TMD) controls have?
At release, the Oretronic IV Tramp Metal Detector (TMD) controls have CE, cCSAus, and RCM approvals. If there are other approvals that are required, please contact the factory at sales.bulk.us@thermofisher.com.
With the Oretronic IV Tramp Metal Detector (TMD) system, can the control be reset remotely when tramp is detected?
No, if tramp is detected, the user needs to check the belt and remove tramp prior to restarting the system.
Is the Oretronic IV Tramp Metal Detector (TMD) system more immune to false trips than the Oretronic III TMD system?
Yes, with factory and field testing, we have proven that the Oretronic IV TMD system is less affected by metal near the system and electrical interference by magnets, motors, and other equipment in the area, minimizing the occurrence of false trips.
Can I connect more than one receiver coil to the Oretronic IV Tramp Metal Detector (TMD) controls?
No. Only one receiver coil can be connected to the Oretronic IV Tramp Metal Detector (TMD) controls.
Why is the Oretronic IV Tramp Metal Detector (TMD) system better than the Oretronic III TMD control?
The Oretronic IV Tramp Metal Detector (TMD) controls offer improved sensitivity, fewer false trips, better user interface, and multi-language software, compared to the Oretronic III TMD control.
Are there special tools required to install an Oretronic IV Tramp Metal Detector (TMD) system?
There are no special tools required to install an Oretronic IV Tramp Metal Detector (TMD) system.
Is the calibration process/menu the same for the Oretronic IV Tramp Metal Detector (TMD) controls as the previous Oretronic III TMD control?
The user interface and calibration menus have changed but the concept of operation and function of fine and coarse sensitivity remains the same.
Is there a stainless steel enclosure option for the Oretronic IV Tramp Metal Detector (TMD) control?
At this time, we do not offer a stainless steel enclosure option for the Oretronic IV Tramp Metal Detector (TMD) control except in Australia. Please contact the factory at sales.bulk.us@thermofisher.com if you have an application which requires a stainless steel enclosure.
Is the Oretronic IV Tramp Metal Detector (TMD) designed for applications in hazardous areas?
The complete system is not designed for applications in hazardous areas. We can provide coils and some accessories suitable for Class II, Division 1 and 2 areas, but the Oretronic IV TMD control must be located outside of the hazardous area. A hazardous area rated junction box can be used to increase the distance at which the control can be mounted from the coils.
What is the detectable tramp size with the Oretronic IV Tramp Metal Detector (TMD) system?
Detectable tramp size depends on several variables including material, burden depth, belt width, belt speed, and coil gap. Please see the sensitivity statement or contact our facility at sales.bulk.us@thermofisher.com to help determine detectable tramp size based on a specific application.
Do the Oretronic IV Tramp Metal Detector (TMD) controls have the same mounting footprint as the previous Oretronic III TMD control?
No, the mounting footprint for the Oretronic IV TMD control versions is not the same as the Oretronic III TMD control. Please refer to the user manual (https://assets.thermofisher.com/TFS-Assets/CAD/manuals/tramp-metal-detector-oretronic-iv-s-manual-REC4479B.pdf) for dimensions and mounting requirements.
What is the difference between the Oretronic IV Tramp Metal Detector, 4A Control and Oretronic IV Tramp Metal Detector, 6A Control?
The Oretronic IV Tramp Metal Detector, 6A Control offers higher power and therefore higher sensitivity to tramp metal detection compared to the Oretronic IV Tramp Metal Detector, 4A Control.
What are the options for the Oretronic IV Tramp Metal Detector (TMD) system?
The Oretronic IV TMD system can be equipped with alarm horn, clip detector, flag drop marker, rotating beacon, high pile detector, RS-485 communication card, and 115 VAC or 230 VAC heater (only for use with metal enclosure, for very cold environments).
Is the Oretronic IV Tramp Metal Detector (TMD) control a direct replacement for the Oretronic III TMD control? Will it work with existing coils?
The Oretronic IV TMD control is a direct replacement for the Oretronic III TMD control and will work with existing Thermo Scientific transmitter, receiver, and under belt coils.
How far from the coils can I mount the Oretronic IV Tramp Metal Detector (TMD) control unit?
The transmitter coil comes with 15 feet of cable and the receiver coil comes with 10 feet of cable. An optional junction box is available to increase the distance by 50 feet.
What are the temperature ratings for non-heated and heated Oretonic IV Tramp Metal Detector (TMD) versions?
Temperature ratings for the non-heated units are -40 to +58 degrees C.
Temperature ratings for the heated units are -50 to +58 degrees C (heater required and only in metal enclosure).
What are the different types of Oretronic IV Tramp Metal Detectors (TMD IV) you offer?
Oretronic IV-4 TMD:
- Improved sensitivity over its predecessor, Oretronic III TMD
- Standard NEMA-4X fiberglass enclosure
- External reset button
- Optional RS-485 communication card
Oretronic IV-6 TMD:
- Higher sensitivity compared to Oretronic IV-4 TMD
- Painted NEMA-4 mild steel metal enclosure
- External reset button
- Optional RS-485 communication card
- Optional heater for low temperature applications, 115V or 230V