PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections
Thermo Scientific™
PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections
Ensure reliable high-pressure connections with biocompatible Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDiameter (English) Inner
P-729Cross0.02 in.
P-704Union0.02 in.
Catalog number P-729
Price (MXN)
-
Type:
Cross
Diameter (English) Inner:
0.02 in.
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections. The material is biocompatible for a wide range of applications.
  • Absolute zero or low dead volume formats
  • Biocompatible
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionPEEK Cross; <0.72 uL Swept Volume
Diameter (English) Inner0.02 in.
For Use With (Equipment)1/16 in. OD Tubing
Length (Metric)27.69 mm
MaterialPEEK
TypeCross
Unit SizeEach