Ethyl Alcohol with USP Grade WFI, 40 proof, GMP
Greenfield Global

Ethyl Alcohol with USP Grade WFI, 40 proof, GMP

This product is made with USP Grade Water for Injection (WFI).
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 gal.
55 gal.
Catalog number P111000040PL05
Price (CLP)
-
Quantity:
5 gal.
This product is made with USP Grade Water for Injection (WFI).
This product is made with USP Grade Water for Injection (WFI). Greenfield Global USA Inc. is not certifying the finished product as injection grade. The suitability of the finished product for injection purposes is the sole responsibility of the end user. This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point87°C to 90°C
CAS64-17-5
Chemical Name or MaterialEthyl Alcohol
ColorColorless
Molecular FormulaC2H6O
PackagingPoly pail
SynonymEthanol, Ethyl Alcohol
GradeUSP, NF
Molecular Weight (g/mol)46.07
Percent Purity19.0–21.0%
Quantity5 gal.
Unit Size5 gal.