Ethyl Alcohol with Deionized Water, 140 proof
Ethyl Alcohol with Deionized Water, 140 proof
Greenfield Global

Ethyl Alcohol with Deionized Water, 140 proof

This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or aRead more
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 gal.
4 x 1 gal.
55 gal.
Catalog number P111000140PL05
Price (USD)
-
Request A Quote
Quantity:
5 gal.
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point87°C to 90°C
CAS64-17-5
Chemical Name or MaterialEthyl Alcohol
ColorColorless
Molecular FormulaC2H6O
PackagingPoly pail
SynonymEthanol, Ethyl Alcohol
Molecular Weight (g/mol)46.07
Percent Purity70%
Quantity5 gal.
Unit Size5 gal.