Ethyl Alcohol, 200 proof, Multi-Compendial, GMP
Greenfield Global

Ethyl Alcohol, 200 proof, Multi-Compendial, GMP

Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 gal.
4 x 4 L
4 x 1 gal.
55 gal.
Catalog number P111WORLD200CS4L
Price (USD)
-
Quantity:
4 x 4 L
Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point87°C to 90°C
CAS64-17-5
Chemical Name or MaterialEthyl Alcohol
ColorColorless
Molecular FormulaC2H6O
PackagingCase
SynonymEthanol, Ethyl Alcohol
GradeMulti-Compendial
Molecular Weight (g/mol)46.07
Percent Purity100%
Quantity4 x 4 L
Unit Size4 x 4 L