P2-CST Multiple Gradient Gel Caster Comb, 25 teeth
Actual product may vary
Thermo Scientific™
P2-CST Multiple Gradient Gel Caster Comb, 25 teeth
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeTeeth CountThickness (Metric) Teeth
P2-20CComb200.8 mm
P2-10CComb100.8 mm
P2-10DComb101.5 mm
P2-15CComb150.8 mm
P2-15DComb151.5 mm
P2-20DComb201.5 mm
P2-20FGFrosted Blank GlassNot ApplicableNot Applicable
P2-20GBlank GlassNot ApplicableNot Applicable
P2-25CComb250.8 mm
P2-CSTCasterNot ApplicableNot Applicable
P2-PREPComb21.5 mm
Catalog number P2-20C
Price (USD)
76.95
Online exclusive
85.50
Save 8.55
Each
Type:
Comb
Teeth Count:
20
Thickness (Metric) Teeth:
0.8 mm
Price (USD)
76.95
Online exclusive
85.50
Save 8.55
Each
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
  • With an external gradient maker, gels can be cast with ease and great reproducibility
  • The design and format of this unit will provide years of service in your lab
Specifications
Teeth Count20
Thickness (Metric) Teeth0.8 mm
For Use With (Equipment)Fisherbrand P2-CST System
TypeComb
Unit SizeEach