Search
| Catalog Number | Quantity |
|---|---|
| P21041 | 1 mg |
The horseradish peroxidase (HRP) conjugate of protein G can be used to detect the Fc portion of various classes and subclasses of immunoglobulins from a variety of species. Protein G is a bacterial protein similar to protein A but with superior selectivity for mouse and human IgG.
For Research Use Only. Not for use in diagnostic procedures.