PiPer™ Pyrophosphate Assay Kit - FAQs
PiPer™ Pyrophosphate Assay Kit - FAQs
View additional product information for PiPer™ Pyrophosphate or Phosphate Assay Kit - FAQs (P22061, P22062)
1 product FAQs found
What is the excitation/emission maxima of resorufin?
The excitation/emission maxima is 563/587 nm.