May I use other fluorophores with cells labeled with pHrodo Deep Red Mammalian and Bacterial Cell Labeling Kit (Cat. No. P35357)?
Yes. The pHrodo Deep Red Mammalian and Bacterial Cell Labeling Kit (Cat. No. P35357) will label the surface of the cells. You may use other fluorophores to stain or label other organelles in fluorescent colors that do not overlap with the far-red signal of pHrodo Deep Red.