May I store pHrodo Deep Red TFP Ester (Cat. Nos. P35358, P35259) labeled antibodies or other proteins in solutions containing sodium azide?
We do not recommend storing antibodies or proteins labeled with pHrodo Deep Red TFP Ester (Cat. Nos. P35358, P35359) in solutions containing sodium azide, as pHrodo Deep Red is not stable in the presence of sodium azide. Consider using an alternative, non-azide-based anti-microbial.