pHrodo™ Red Avidin (Fluorogenic pH Sensor)
Invitrogen™
pHrodo™ Red Avidin (Fluorogenic pH Sensor)
pHrodo™ Red Avidin is a conjugate of avidin and pHrodo™ Red dye, a novel, fluorogenic dye that dramatically increases inRead more
Have Questions?
Catalog NumberQuantity
P353621 mg
Catalog number P35362
Price (USD)
696.00
Each
Quantity:
1 mg
Price (USD)
696.00
Each
pHrodo™ Red Avidin is a conjugate of avidin and pHrodo™ Red dye, a novel, fluorogenic dye that dramatically increases in fluorescence as the pH of its surroundings become more acidic. pHrodo™ Red dye conjugates are non-fluorescent outside the cell, but fluoresce bright red in phagosomes, making them ideal tools for studies ranging from phagocytosis of bioparticles to receptor internalization. Original pHrodo™ and pHrodo™ Red dye are the same molecule.
With pHrodo™ Red Avidin Fluorogenic pH Sensor You Get:
• Specific detection of phagocytosis and endocytosis
• Faster staining
• More accurate results in sensitive experiments
• Multiplexing capability with green dyes
• Validation in flow cytometry, imaging, HTS, and HCS
Get Better Results in Endocytosis Studies
Endocytosis and phagocytosis underly physiological events ranging from host-pathogen response to receptor desensitization via internalization as is common in the GPCR class of receptors). Once internalized, material can be acidified in endosomes or lysosomes. Our pHrodo™ Red dye fluorogenic pH sensitive probe, when conjugated to target molecules, allows direct visual monitoring of material entering these acidic compartments. At neutral and basic pH values, pHrodo™ Red dye emission is at its minimum; with acidification, pHrodo™ Red dye emission is increased by as much as 8-fold (at pH 4). The red fluorescence of pHrodo™ Red Avidin is ideal for multiplexing applications with green-fluorescent compounds such GFP, Fluo-4, or calcein.
Faster and More Accurate Staining in Multiple Validated Applications
pHrodo™ Red Avidin offers reduced signal variability and improved timing compared to ratiometric dyes like BCECF and SNARF, and the lack of fluorescence outside the cell eliminates the need for wash steps and quencher dyes. The fluorescence of pHrodo™ Red dye is dimmed by complete media and whole blood, further improving the signal-to-noise ratio. pHrodo™ Red has been cited in over 150 publications and is validated in flow cytometry, high throughput screening (HTS), high content screening (HCS), and other imaging applications.
Easy Biotinylation Lets You Enjoy Flexibility in Experimental Design
The pHrodo™ Red Avidin reagent can quickly be coupled to any biotinlyated reagent, including many of our own biotin conjugates of antibodies, proteins, and toxins including transferrin, EGF, and cholera toxin. Alternatively, you can create your own biotinylated target either by using Click-iT™ chemistry to metabolically incorporate an alkyne biotin, or by using any of our biotin labeling kits.
Alternative Conjugation Chemistry to pHrodo™ SE
In cases where the amine reactive pHrodo-SE conjugates show reduced or complete loss of function, biotin and avidin conjugation can be considered using pHrodo™ Red Avidin. In one example, pHrodo-EGF was created using the avidin-biotin approach and shown to bind its receptor and be internalized (Ref 1).
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Reference:
1. Suprynowicz FA, Krawczyk E, Hebert JD, Sudarshan SR, Simic V, Kamonjoh CM, Schlegel R. The human papillomavirus type 16 E5 oncoprotein inhibits epidermal growth factor trafficking independently of endosome acidification. J Virol. 2010 Oct;84(20):10619-29. PubMed PMID: 20686024
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeAvidin
Quantity1 mg
Shipping ConditionRoom Temperature
ConjugatepHrodo Red
Product LineMolecular Probes, pHrodo
Unit SizeEach
Contents & Storage
Store at ≤-20°C
Desiccate and protect from light
Frequently asked questions (FAQs)
I am performing a phagocytosis assay of macrophages engulfing pHrodo-labeled bacteria. What do you recommend for fixation after the phagocytosis?
pHrodo is relatively non-fluorescent until it enters the acidic phagosome, at which point its fluorescence increases. If you fix the sample, the pHrodo will only reflect the pH of the buffer the cells are in, and not the pH of the phagosome. For this reason, we do not recommend fixing samples. If you want to see how many cells engulfed the labeled bacteria, fix the cells and then place the fixed cells in an acidic buffer for the assay.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Citations & References (2)
Citations & References
Abstract
Simultaneous TCR and CD244 Signals Induce Dynamic Downmodulation of CD244 on Human Antiviral T Cells.
Authors:Pacheco Y, McLean AP, Rohrbach J, Porichis F, Kaufmann DE, Kavanagh DG,
Journal:
PubMed ID:23913963
'Various cosignaling molecules on T cells can contribute to activation, inhibition, or exhaustion, depending on context. The surface receptor signaling lymphocytic activation molecule (SLAM) family receptor CD244 (2B4/SLAMf4) has been shown to be capable of either inhibitory or enhancing effects upon engagement of its ligand CD48 (SLAMf2). We examined phenotypes ... More
Intracellular uptake of macromolecules by brain lymphatic endothelial cells during zebrafish embryonic development.
Authors:van Lessen M, Shibata-Germanos S, van Impel A, Hawkins TA, Rihel J, Schulte-Merker S
Journal:Elife
PubMed ID:28498105
'The lymphatic system controls fluid homeostasis and the clearance of macromolecules from interstitial compartments. In mammals brain lymphatics were only recently discovered, with significant implications for physiology and disease. We examined zebrafish for the presence of brain lymphatics and found loosely connected endothelial cells with lymphatic molecular signature covering parts ... More