pHrodo™ Red and Green Epidermal Growth Factor (EGF) and pHrodo™ Red Transferrin Conjugates
pHrodo™ Red and Green Epidermal Growth Factor (EGF) and pHrodo™ Red Transferrin Conjugates
pHrodo™ Red and Green Epidermal Growth Factor (EGF) and pHrodo™ Red Transferrin Conjugates
Invitrogen™

pHrodo™ Red and Green Epidermal Growth Factor (EGF) and pHrodo™ Red Transferrin Conjugates

Detect internalized EGF and transferrin in cells with the pHrodo Red and pHrodo Green Epidermal Growth Factor (EGF) and pHrodo Red Transferrin conjugates, for use in applications such as HCS, flow cytometry, microplate-based fluorometry, and HTS.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
P35375Green EGF Conjugate
P35376Red Transferrin Conjugate
P35374Red EGF Conjugate
Catalog number P35375
Price (USD)
598.00
Each
Product Type:
Green EGF Conjugate
Price (USD)
598.00
Each
Detect internalized EGF or transferrin with highly active, pH-sensitive pHrodo Red and pHrodo Green Epidermal Growth Factor (EGF) conjugates, and the pHrodo Red Transferrin Conjugate. These conjugates consist of pHrodo dyes directly linked to highly active EGF or transferrin molecules, which can be used in applications including HCS, flow cytometry, microplate fluorometry, and HTS to measure internalized EGF and transferrin molecules. The pHrodo Red and pHrodo Green dyes fluoresce brightly when exposed to acidic environments, such as those found in endosomes and lysosomes.

The pHrodo Red and pHrodo Green Epidermal Growth Factor (EGF) conjugates consist of pHrodo Red or pHrodo Green dye directly conjugated to a highly active human recombinant EGF molecule and are used for the detection of internalized EGF receptor in cells. Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more. Fluorescent labels that respond to a pH change add another level of detection.

pHrodo Red Transferrin Conjugate consists of pHrodo Red dye directly conjugated to a highly active transferrin molecule and is used for the detection of internalized transferrin in cells. Transferrin is a monomeric serum glycoprotein (∼80,000 daltons) that binds to a specific receptor on the surface of vertebrate cells and delivers up to two Fe3+ atoms via receptor-mediated endocytosis. Once iron-carrying transferrin proteins are inside endosomes, the acidic environment favors dissociation of iron from the transferrin–receptor complex. Following the release of iron, the apotransferrin is recycled to the plasma membrane, where it is released from its receptor to scavenge more iron.

The highly photostable pHrodo Red and pHrodo Green dyes are weakly fluorescent outside of cells at neutral pH, but when internalized specifically through EGF or transferrin endocytosis, become brightly fluorescent in acidic endosomes. This allows the measurement of internalized EGF or transferrin, as well as the dynamic monitoring of EGF or transferrin internalization in live cells.

The pHrodo Red EGF Conjugate and the pHrodo Red Transferrin Conjugate have excitation/emission spectra of 560/585 nm and can be multiplexed with a wide variety of blue, red, and far-red dyes reporters such as Mitosox Red, CellEvent Caspase 3/7 Red, NucBlue, RFPs, and Mitotracker Deep Red, among many others. The pHrodo Green EGF Conjugate has excitation/emission spectra of 509/533 nm and can be multiplexed with a wide variety of blue, red, and far-red dyes reporters such as Mitosox Red, CellEvent Caspase 3/7 Red, NucBlue, RFPs, and Mitotracker Deep Red, among many others.

The pHrodo Red and pHrodo Green EGF conjugates and the pHrodo Red Transferrin Conjugate are compatible in applications including traditional fluorescence microscopy, high content screening (HCS), flow cytometry, and microplate-based fluorimetry or high throughput screening (HTS). The conjugates are also compatible with various widefield or confocal fluorescence microscopes, flow cytometers, fluorescence plate readers, and high content instruments.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionpHrodo™ Green Epidermal Growth Factor (EGF) Conjugate
Detection MethodFluorescence
Excitation/Emission509/533 nm
FormLyophilized Powder
Protein FamilyGrowth Factors and Receptors
Quantity20 μg
Sub Cellular LocalizationEndosomes, Lysosomes
ColorGreen
For Use With (Application)Cell Analysis
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer, Fluorometer, High Content Analysis Instrument
Product LinepHrodo
Product TypeGreen EGF Conjugate
Unit SizeEach
Contents & Storage
Store at -5° to -30°C, desiccate, and protect from light.

Citations & References (4)

Citations & References
Abstract
The human papillomavirus type 16 E5 oncoprotein inhibits epidermal growth factor trafficking independently of endosome acidification.
Authors:Suprynowicz FA, Krawczyk E, Hebert JD, Sudarshan SR, Simic V, Kamonjoh CM, Schlegel R,
Journal:J Virol
PubMed ID:20686024
The human papillomavirus type 16 E5 oncoprotein (16E5) enhances acute, ligand-dependent activation of the epidermal growth factor receptor (EGFR) and concomitantly alkalinizes endosomes, presumably by binding to the 16-kDa  ... More
A novel TIP30 protein complex regulates EGF receptor signaling and endocytic degradation.
Authors:Zhang C, Li A, Zhang X, Xiao H,
Journal:J Biol Chem
PubMed ID:21252234
Activated epidermal growth factor receptor (EGFR) continues to signal in the early endosome, but how this signaling process is regulated is less well understood. Here we describe a protein complex consisting of TIP30, endophilin B1, and acyl-CoA synthetase long chain family member 4 (ACSL4) that interacts with Rab5a and regulates ... More
EWI-2 controls nucleocytoplasmic shuttling of EGFR signaling molecules and miRNA sorting in exosomes to inhibit prostate cancer cell metastasis.
Authors:
Journal:Mol Oncol
PubMed ID:33605506
Culture and Nucleofection of Postnatal Day 7 Cortical and Cerebellar Mouse Astroglial Cells.
Authors:
Journal:Bio Protoc
PubMed ID:34179254