Owl™ P7-CST Multiple Gradient Gel Caster
Actual product may vary
Thermo Scientific™
Owl™ P7-CST Multiple Gradient Gel Caster
Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
P7-SCSpacer, Thin
P7-10FGFrosted Blank Glass
P7-10FRFrosted Notched Glass
P7-10GBlank Glass
P7-10RNotched Glass
P7-10RANotched Alumina Plate
P7-CSTCaster
P7-SDSpacer, Thick
Catalog number P7-SC
Price (USD)
28.26
Online exclusive
31.40
Save 3.14
Pack of
Type:
Spacer, Thin
Price (USD)
28.26
Online exclusive
31.40
Save 3.14
Pack of
Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster. One of the simplest devices to cast gradient gels, this caster is compatible with the Owl™ PBDS system.
  • With an external gradient maker, gels can be cast with ease and great reproducibility
  • The design and format of this unit will provide years of service in your lab
Specifications
DescriptionSpacer (2 sides,1 bottom)
Thickness (Metric)0.8 mm
Width (Metric)0.5 cm
For Use With (Equipment)Owl P7-CST System
Product LineOwl
TypeSpacer, Thin
Unit SizePack of