PS-Speck™ Microscope Point Source Kit (blue, green, orange & deep-red fluorescent beads) - FAQs
PS-Speck™ Microscope Point Source Kit (blue, green, orange & deep-red fluorescent beads) - FAQs
View additional product information for PS-Speck™ Microscope Point Source Kit (blue, green, orange & deep-red fluorescent beads) - FAQs (P7220)
1 product FAQs found
What is the refractive index of the mounting medium in PS-Speck Microscope Point Source Kit (Cat. No. P7220)?
The mountant (Component E) has a refractive index of ~1.47.