Owl™ P81 Single-Sided Vertical Electrophoresis System, 10 x 10cm gel size, 150mL buffer volume
Perform quick protein runs with the economical and simple, yet rugged Thermo Scientific™ Owl™ P81 Single-Sided Vertical Electrophoresis System.
Have Questions?
Catalog Number
Length (Metric) Gel
P81
10 cm
Catalog number P81
Price (USD)
831.60
Online exclusive
924.00
Save 92.40
Each
Length (Metric) Gel:
10 cm
Price (USD)
831.60
Online exclusive
924.00
Save 92.40
Each
Perform quick protein runs with the economical and simple, yet rugged Thermo Scientific™ Owl™ P81 Single-Sided Vertical Electrophoresis System.
Features:
Upper buffer chamber extends the length of the gel to distribute heat evenly across the entire gel surface
Side clamps press glass plates against silicone gasket to provide uniform pressure and form a leakproof seal resulting in flat, even banding patterns
Notched alumina ceramic plate (included) may be used in front of notched glass for cooler, faster runs
Includes:
Upper and lower buffer chambers
Thermo Scientific™ SuperSafe™ lid with attached power supply leads
Blank glass plate
Notched glass plate
Notched alumina plate
Two combs (10 well, 0.8mm thick)
Spacer set (0.8mm thick)
Specifications
Certifications/ComplianceCE marked
DescriptionGel size: 10 x 10cm; Buffer volume: 150mL; L x W x H: 18 x 8 x 13cm
Dimensions (L x W x H)7.1 x 3.1 x 5.1 in. (18 x 8 x 13 cm)
Height (English)5.1 in.
Height (Metric)13 cm
Length (English)7.1 in.
Length (Metric)18 cm
Length (Metric) Gel10 cm
Volume (Metric) Buffer150 mL
Width (English)3.1 in.
Width (Metric)8 cm
Width (Metric) Gel10 cm
Product LineOwl
TypeSingle-Sided Vertical Electrophoresis System
Unit SizeEach
Frequently asked questions (FAQs)
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.