R-Phycoerythrin, Biotin-XX Conjugate
Invitrogen™

R-Phycoerythrin, Biotin-XX Conjugate

R-Phycoerythrin, Biotin-XX Conjugate
Have Questions?
Catalog NumberQuantity
P811
also known as P-811
0.5 mL
Catalog number P811
also known as P-811
Price (USD)
453.00
Each
Quantity:
0.5 mL
Price (USD)
453.00
Each

R-phycoerythrin biotin-XX conjugate consists of the exceptionally bright phycobiliprotein, R-PE conjugated with biotin-XX, where the biotin is well separated from the point of attachment by a fourteen-atom spacer.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Concentration4 mg/mL
Product TypeBiotin Conjugate (R-PE)
Protein FamilyOther Proteins
Quantity0.5 mL
Shipping ConditionWet Ice
ConjugateBiotin
FormLiquid
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Citations & References (5)

Citations & References
Abstract
Post-column reaction detection based on fluorescence energy transfer in the far red spectral region.
Authors:Shahdeo K,Karnes HT
Journal:Journal of pharmaceutical and biomedical analysis
PubMed ID:10703992
Simultaneous multicolor fluorescence cross-correlation spectroscopy to detect higher order molecular interactions using single wavelength laser excitation.
Authors:Hwang LC, Gösch M, Lasser T, Wohland T
Journal:Biophys J
PubMed ID:16632502
'Fluorescence cross-correlation spectroscopy is a powerful method for the study of molecular interactions and dynamics in solution and even in living cells. Usually, in the optical setup, either two laser beams have to be superimposed in their respective confocal volumes or two-photon excitation is used for a dual-color detection system. ... More
P-selectin glycoprotein ligand-1 supports rolling on E- and P-selectin in vivo.
Authors:Norman KE, Katopodis AG, Thoma G, Kolbinger F, Hicks AE, Cotter MJ, Pockley AG, Hellewell PG
Journal:Blood
PubMed ID:11071658
'Selectin-dependent rolling is the earliest observable event in the recruitment of leukocytes to inflamed tissues. Several glycoproteins decorated with sialic acid, fucose, and/or sulfate have been shown to bind the selectins. The best-characterized selectin ligand is P-selectin glycoprotein-1 (PSGL-1) that supports P-selectin- dependent rolling in vitro and in vivo. In ... More
The effect of IL-2 treatment on transcriptional attenuation in proto-oncogenes pim-1 and c-myb in human thymic blast cells.
Authors:Rohwer F, Todd S, McGuire KL
Journal:J Immunol
PubMed ID:8752912
IL-2 is the major mitogenic cytokine for mature human T cells. This growth factor has been shown previously to induce the expression of a number of genes, including structural proteins, proto-oncogenes, and metabolic enzymes. Multiple mechanisms, including increases in mRNA stability, protein synthesis, and new transcriptional initiation, have been studied ... More
A TAT-streptavidin fusion protein directs uptake of biotinylated cargo into mammalian cells.
Authors:Albarran B, To R, Stayton PS
Journal:Protein Eng Des Sel
PubMed ID:15820981
The HIV-1 TAT peptide has been used extensively for directing the intracellular delivery of an assortment of cargo, including DNA, liposomes and macromolecules. For protein delivery, a variety of TAT-fusion proteins have been described which link the TAT coding sequence to the protein coding sequence of interest. Streptavidin represents a ... More