Owl™ P82 Dual-Gel Electrophoresis Double Sided Vertical System, 8-10 x 10cm gel, 150-300mL buffer volume
Thermo Scientific™

Owl™ P82 Dual-Gel Electrophoresis Double Sided Vertical System, 8-10 x 10cm gel, 150-300mL buffer volume

Produce flat, even banding and crisp resolution with the easy-to-use Thermo Scientific™ Owl™ P82 Dual-Gel Electrophoresis System.
Have Questions?
Catalog NumberLength (Metric) Gel
P828 to 10 cm
Catalog number P82
Price (USD)
1,368.00
Online exclusive
1,520.00
Save 152.00
Each
Length (Metric) Gel:
8 to 10 cm
Price (USD)
1,368.00
Online exclusive
1,520.00
Save 152.00
Each
Produce flat, even banding and crisp resolution with the easy-to-use Thermo Scientific™ Owl™ P82 Dual-Gel Electrophoresis System.
Features:
  • Runs two 10 x 10cm gels or one gel when used with blocking plate; accommodates 8 x 10cm gels utilizing the provided adapter
  • Allows gels to easily be placed into the device
  • Wedge placed in front of the cassettes provides even pressure against the leakproof gasket, places the cassettes in proper running position
  • Two sizes of wedges accommodate varying thickness of precast gels
  • Flexible—use precast polyacrylamide gels or hand cast gels
  • No cooling required

Includes:

  • Inner buffer chambers
  • Buffer tank
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Two wedges (1.27cm thick)
  • Two wedges (1.59cm thick)
  • Adapter plates for 10 × 8 gels (2)
  • Blocking plate
Specifications
Certifications/ComplianceCE marked
DescriptionDouble Sided Vertical System, 8-10 x 10cm gel system, 150mL to 300mL buffer volume
Dimensions (L x W x H)5.88 x 4.75 x 6.69 in. (15.2 x 12.4 x 17 cm)
Height (English)6.69 in.
Height (Metric)17 cm
Length (English)5.88 in.
Length (Metric)15.2 cm
Length (Metric) Gel8 to 10 cm
Max. Current15 mA
Volume (Metric) Lower Buffer Chamber Max.300 mL
Volume (Metric) Lower Buffer Chamber Min.150 mL
Volume (Metric) Upper Buffer Chamber150 mL
Width (English)4.75 in.
Width (Metric)12.4 cm
Width (Metric) Gel10 cm
Max. Voltage330 V
Product LineOwl
TypeDual-Gel Electrophoresis Double Sided Vertical System
Unit SizeEach

Frequently asked questions (FAQs)

Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?

The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.